Assay Details
Gene Symbol: TLR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135928.1 | NP_001129400.1 | 4 - 5 | 2554 | 97 | |
XM_012105647.3 | XP_011961037.2 | 4 - 5 | 2878 | 97 | ||
GenBank mRNA | AM231299.1 | - | 1 - 2 | 171 | 97 | |
AM981301.1 | - | 4 - 5 | 2554 | 97 | ||
AY957614.1 | - | 1 - 2 | 100 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
