Assay Details
| Gene Symbol: EIF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162563.1 | NP_001156035.1 | 4 - 5 | 548 | 64 | |
| XM_012188071.3 | XP_012043461.1 | 5 - 6 | 665 | 64 | ||
| XM_012188072.2 | XP_012043462.1 | 5 - 6 | 664 | 64 | ||
| GenBank mRNA | FJ958353.1 | - | 4 - 5 | 548 | 64 | |
More Information
Set Membership: |
|

EIF6
Genomic Map