Assay Details
Gene Symbol: PUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163051.1 | NP_001156523.1 | 1 - 2 | 226 | 97 | |
XM_012097268.3 | XP_011952658.2 | 2 - 3 | 725 | 97 | ||
XM_012097269.3 | XP_011952659.2 | 2 - 3 | 286 | 97 | ||
GenBank mRNA | FJ958366.1 | - | 1 - 2 | 226 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
