Assay Details
| Gene Symbol: TTPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198882.1 | NP_001185811.1 | 2 - 3 | 375 | 75 | |
| GenBank mRNA | HQ268507.1 | - | 2 - 3 | 375 | 75 | |
More Information
Set Membership: |
|

TTPA
Genomic Map