Assay Details
| Gene Symbol: IDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009276.1 | NP_001009276.1 | 8 - 9 | 1162 | 62 | |
| XM_027963886.1 | XP_027819687.1 | 8 - 9 | 1312 | 62 | ||
| GenBank mRNA | AY208678.1 | - | 8 - 9 | 1162 | 62 | |
| DQ239637.1 | - | 1 - 2 | 41 | 62 | ||
More Information
Set Membership: |
|

IDH1
Genomic Map