Assay Details
| Gene Symbol: ARF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162545.1 | NP_001156017.1 | 3 - 4 | 387 | 62 | |
| XM_012158906.3 | XP_012014296.1 | 4 - 5 | 701 | 62 | ||
| GenBank mRNA | FJ969413.1 | - | 3 - 4 | 387 | 62 | |
More Information
Set Membership: |
|

ARF3
Genomic Map