Assay Details
| Gene Symbol: SLC17A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009742.1 | NP_001009742.1 | 3 - 4 | 606 | 60 | |
| XM_015097251.2 | XP_014952737.1 | 3 - 4 | 1595 | 60 | ||
| GenBank mRNA | AF244578.1 | - | 3 - 4 | 606 | 60 | |
More Information
Set Membership: |
|

SLC17A5
Genomic Map