Assay Details
| Gene Symbol: EIF1AX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145179.1 | NP_001138651.1 | 5 - 6 | 514 | 71 | |
| GenBank mRNA | FJ546081.1 | - | 5 - 6 | 514 | 71 | |
More Information
Set Membership: |
|

EIF1AX
Genomic Map