Assay Details
| Gene Symbol: TNFRSF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166185.1 | NP_001159657.1 | 5 - 6 | 545 | 86 | |
| GenBank mRNA | GQ221036.1 | - | 5 - 6 | 545 | 86 | |
More Information
Set Membership: |
|

TNFRSF1A
Genomic Map