Assay Details
| Gene Symbol: ATP5PD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142891.1 | NP_001136363.1 | 5 - 6 | 390 | 88 | |
| GenBank mRNA | DQ377365.1 | - | 5 - 6 | 390 | 88 | |
More Information
Set Membership: |
|

ATP5PD
Genomic Map