Assay Details
| Gene Symbol: UCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001280694.1 | NP_001267623.1 | 1 - 2 | 350 | 55 | |
| GenBank mRNA | JN604984.1 | - | 1 - 2 | 350 | 55 | |
More Information
Set Membership: |
|

UCP1
Genomic Map