Assay Details
| Gene Symbol: FGF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009230.1 | NP_001009230.1 | 2 - 3 | 1835 | 110 | |
| GenBank mRNA | AF213396.1 | - | 2 - 3 | 1835 | 110 | |
More Information
Set Membership: |
|

FGF10
Genomic Map