Assay Details
Gene Symbol: AK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161882.1 | NP_001155354.1 | 4 - 5 | 519 | 62 | |
GenBank mRNA | FJ943993.1 | - | 4 - 5 | 519 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
