Assay Details
| Gene Symbol: KRTAP6-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193399.1 | NP_001180328.1 | 1 | 127 | 91 | |
| GenBank mRNA | HM066201.1 | - | 1 | 93 | 91 | |
| JQ177212.1 | - | 1 | 98 | 91 | ||
More Information
Set Membership: |
|

KRTAP6-1
Genomic Map