Assay Details
Gene Symbol: LOC101109747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280717.1 | NP_001267646.1 | 1 - 2 | 75 | 108 | |
XM_012100429.3 | XP_011955819.2 | 1 - 2 | 163 | 108 | ||
GenBank mRNA | FM986335.1 | - | 1 - 2 | 78 | 108 | |
FM986336.1 | - | 1 - 2 | 78 | 108 | ||
JF735994.1 | - | 1 - 2 | 75 | 108 | ||
JQ824376.1 | - | 1 - 2 | 103 | 108 | ||
M73983.1 | - | 1 - 2 | 100 | 108 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
