Assay Details
Gene Symbol: GNRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009397.1 | NP_001009397.1 | 2 - 3 | 794 | 107 | |
XM_012179358.3 | XP_012034748.1 | 2 - 3 | 1763 | 107 | ||
GenBank mRNA | L22215.1 | - | 2 - 3 | 781 | 107 | |
X72088.1 | - | 794 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
