Assay Details
Gene Symbol: CA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009408.2 | NP_001009408.1 | 7 - 8 | 1035 | 97 | |
GenBank mRNA | Y07785.1 | - | 1036 | 97 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009408.2 | NP_001009408.1 | 7 - 8 | 1035 | 97 | |
GenBank mRNA | Y07785.1 | - | 1036 | 97 |