Assay Details
| Gene Symbol: NFIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142892.1 | NP_001136364.1 | 1 | 1318 | 127 | |
| XM_012110388.2 | XP_011965778.1 | 2 | 2406 | 127 | ||
| XM_015092735.2 | XP_014948221.1 | 2 | 1609 | 127 | ||
| XM_015092737.2 | XP_014948223.1 | 3 | 3786 | 127 | ||
| GenBank mRNA | EU293838.1 | - | 1 | 1318 | 127 | |
More Information
Set Membership: |
|

NFIL3
Genomic Map