Assay Details
Gene Symbol: ARL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162553.2 | NP_001156025.1 | 4 - 5 | 331 | 90 | |
GenBank mRNA | EE773754.1 | - | 3 - 4 | 319 | 90 | |
EE834734.1 | - | 4 - 5 | 364 | 90 | ||
FJ969414.1 | - | 3 - 4 | 312 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
