Assay Details
Gene Symbol: RACK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281479.1 | NP_001268408.1 | 2 - 3 | 349 | 69 | |
GenBank mRNA | KC422451.1 | - | 2 - 3 | 349 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
