Assay Details
| Gene Symbol: ADCYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009776.1 | NP_001009776.1 | 4 - 5 | 918 | 74 | |
| GenBank mRNA | S83511.1 | - | 4 - 5 | 918 | 74 | |
More Information
Set Membership: |
|

ADCYAP1
Genomic Map