Assay Details
Gene Symbol: ITM2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131031.1 | NP_001124503.1 | 3 - 4 | 468 | 65 | |
GenBank mRNA | EU797608.1 | - | 3 - 4 | 468 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
