Assay Details
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177679.1 | NP_001171150.1 | 2 - 3 | 208 | 71 | |
GenBank mRNA | HM012713.1 | - | 2 - 3 | 208 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
