Assay Details
Gene Symbol: MC5R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078656.1 | NP_001072124.1 | 1 | 917 | 68 | |
GenBank mRNA | DQ530056.1 | - | 1 | 917 | 68 | |
DQ530057.1 | - | 1 | 917 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
