Assay Details
| Gene Symbol: KLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093787.1 | NP_001087256.1 | 1 - 2 | 46 | 70 | |
| GenBank mRNA | EF521403.1 | - | 1 - 2 | 46 | 70 | |
More Information
Set Membership: |
|

KLK1
Genomic Map