Assay Details
| Gene Symbol: PSMA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139446.2 | NP_001132918.1 | 2 - 3 | 261 | 82 | |
| GenBank mRNA | FJ223570.1 | - | 2 - 3 | 262 | 82 | |
More Information
Set Membership: |
|

PSMA6
Genomic Map