Assay Details
Gene Symbol: QPCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166178.1 | NP_001159650.1 | 5 - 6 | 820 | 81 | |
GenBank mRNA | AF039441.1 | - | 4 - 5 | 606 | 81 | |
GQ221058.1 | - | 5 - 6 | 820 | 81 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
