Assay Details
| Gene Symbol: BRMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166189.1 | NP_001159661.1 | 2 - 3 | 231 | 57 | |
| XM_027959245.1 | XP_027815046.1 | 3 - 4 | 373 | 57 | ||
| XM_027959246.1 | XP_027815047.1 | 3 - 4 | 534 | 57 | ||
| XM_027959247.1 | XP_027815048.1 | 3 - 4 | 531 | 57 | ||
| GenBank mRNA | GQ221051.1 | - | 2 - 3 | 231 | 57 | |
More Information
Set Membership: |
|

BRMS1
Genomic Map