Assay Details
| Gene Symbol: UBA52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009286.1 | NP_001009286.1 | 3 | 221 | 104 | |
| XM_012177581.2 | XP_012032971.1 | 4 | 335 | 104 | ||
| GenBank mRNA | AY563026.1 | - | 3 | 221 | 104 | |
More Information
Set Membership: |
|

UBA52
Genomic Map