Assay Details
Gene Symbol: C4H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007949.3 | XP_004007998.2 | 6 - 7 | 717 | 85 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C4H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007949.3 | XP_004007998.2 | 6 - 7 | 717 | 85 |