Assay Details
Gene Symbol: GPATCH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006022.4 | XP_004006071.2 | 8 - 9 | 894 | 77 | |
XM_012173551.3 | XP_012028941.2 | 9 - 10 | 1011 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
