Assay Details
| Gene Symbol: ZC2HC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011754.4 | XP_004011803.1 | 7 - 8 | 783 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZC2HC1A| Gene Symbol: ZC2HC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011754.4 | XP_004011803.1 | 7 - 8 | 783 | 64 | |
Set Membership: |
|