Assay Details
| Gene Symbol: LSM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018551.4 | XP_004018600.1 | 3 - 4 | 316 | 105 | |
| Gene Symbol: LOC114111228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962934.1 | XP_027818735.1 | 1 | 268 | 105 | |
| Gene Symbol: LOC114114607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969111.1 | XP_027824912.1 | 1 | 247 | 105 | |
More Information
Set Membership: |
|

LSM3
Genomic Map