Assay Details
Gene Symbol: MLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959144.1 | XP_027814945.1 | 8 - 9 | 2523 | 61 | |
XM_027959145.1 | XP_027814946.1 | 8 - 9 | 2523 | 61 | ||
XM_027959146.1 | XP_027814947.1 | 8 - 9 | 2523 | 61 | ||
XM_027959147.1 | XP_027814948.1 | 8 - 9 | 2523 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
