Assay Details
| Gene Symbol: SS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012103457.3 | XP_011958847.2 | 5 - 6 | 611 | 60 | |
| XM_015103616.2 | XP_014959102.2 | 5 - 6 | 647 | 60 | ||
| XM_015103617.2 | XP_014959103.2 | 5 - 6 | 647 | 60 | ||
More Information
Set Membership: |
|

SS18
Genomic Map