Assay Details
Gene Symbol: NSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021782.4 | XP_004021831.1 | 22 - 23 | 4414 | 91 | |
XM_012105930.3 | XP_011961320.1 | 22 - 23 | 4381 | 91 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021782.4 | XP_004021831.1 | 22 - 23 | 4414 | 91 | |
XM_012105930.3 | XP_011961320.1 | 22 - 23 | 4381 | 91 |