Assay Details
Gene Symbol: NSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021782.4 | XP_004021831.1 | 9 - 10 | 2406 | 85 | |
XM_012105930.3 | XP_011961320.1 | 9 - 10 | 2406 | 85 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021782.4 | XP_004021831.1 | 9 - 10 | 2406 | 85 | |
XM_012105930.3 | XP_011961320.1 | 9 - 10 | 2406 | 85 |