Assay Details
| Gene Symbol: COL8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002871.3 | XP_004002920.2 | 3 - 4 | 623 | 65 | |
| XM_027979258.1 | XP_027835059.1 | 3 - 4 | 769 | 65 | ||
More Information
Set Membership: |
|

COL8A1
Genomic Map