Assay Details
| Gene Symbol: LOC101119839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976439.1 | XP_027832240.1 | 1 - 2 | 75 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101119839| Gene Symbol: LOC101119839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976439.1 | XP_027832240.1 | 1 - 2 | 75 | 102 | |
Set Membership: |
|