Assay Details
Gene Symbol: ATP6V1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006972.3 | XP_004007021.1 | 8 - 9 | 749 | 57 | |
GenBank mRNA | EU370552.1 | - | 5 - 6 | 364 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
