Assay Details
| Gene Symbol: LOC101121602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012178613.3 | XP_012034003.2 | 28 - 29 | 5171 | 98 | |
| XM_012178614.3 | XP_012034004.2 | 27 - 28 | 5077 | 98 | ||
| XM_027970311.1 | XP_027826112.1 | 28 - 29 | 5171 | 98 | ||
| XM_027970312.1 | XP_027826113.1 | 27 - 28 | 5114 | 98 | ||
More Information
Set Membership: |
|

LOC101121602
Genomic Map