Assay Details
Gene Symbol: SPG21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010224.4 | XP_004010273.1 | 3 - 4 | 396 | 83 | |
XM_012180730.3 | XP_012036120.1 | 3 - 4 | 349 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
