Assay Details
Gene Symbol: CADM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016040.4 | XP_004016089.2 | 1 - 2 | 145 | 67 | |
XM_012095482.3 | XP_011950872.2 | 1 - 2 | 151 | 67 | ||
XM_012095483.3 | XP_011950873.2 | 1 - 2 | 151 | 67 | ||
XM_012095484.3 | XP_011950874.2 | 1 - 2 | 151 | 67 | ||
XM_012095485.3 | XP_011950875.2 | 1 - 2 | 136 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
