Assay Details
| Gene Symbol: TSC22D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022438.4 | XP_004022487.1 | 2 - 3 | 483 | 76 | |
| XM_004022439.4 | XP_004022488.1 | 2 - 3 | 143 | 76 | ||
| XM_027963356.1 | XP_027819157.1 | 3 - 4 | 638 | 76 | ||
| XM_027963357.1 | XP_027819158.1 | 3 - 4 | 671 | 76 | ||
| XM_027963358.1 | XP_027819159.1 | 2 - 3 | 435 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004022439.4 transcripts.
Set Membership: |
|

TSC22D3
Genomic Map