Assay Details
Gene Symbol: ELOF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008548.4 | XP_004008597.1 | 2 - 3 | 329 | 62 | |
XM_012177938.3 | XP_012033328.1 | 2 - 3 | 161 | 62 | ||
XM_027969699.1 | XP_027825500.1 | 2 - 3 | 236 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
