Assay Details
| Gene Symbol: LOC114116372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973017.1 | XP_027828818.1 | 1 - 2 | 381 | 67 | |
| XM_027973018.1 | XP_027828819.1 | 1 - 2 | 384 | 67 | ||
| Gene Symbol: LOC101103244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973014.1 | XP_027828815.1 | 26 - 27 | 3575 | 67 | |
More Information
Set Membership: |
|

LOC114116372
Genomic Map