Assay Details
Gene Symbol: LOC101112784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020955.4 | XP_004021004.1 | 14 - 15 | 1401 | 94 | |
XM_027962096.1 | XP_027817897.1 | 14 - 15 | 1589 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
