Assay Details
Gene Symbol: LOC114112698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963847.1 | XP_027819648.1 | 3 - 4 | 208 | 86 |
Gene Symbol: EXOC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012173630.2 | XP_012029020.2 | 5 - 6 | 597 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
