Assay Details
| Gene Symbol: RSPH10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021053.4 | XP_004021102.1 | 13 - 14 | 1774 | 60 | |
| XM_027961776.1 | XP_027817577.1 | 13 - 14 | 1774 | 60 | ||
| XM_027961777.1 | XP_027817578.1 | 13 - 14 | 1774 | 60 | ||
More Information
Set Membership: |
|

RSPH10B
Genomic Map