Assay Details
Gene Symbol: MFSD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095897.2 | XP_014951383.2 | 4 - 5 | 953 | 76 | |
XM_027969884.1 | XP_027825685.1 | 4 - 5 | 955 | 76 | ||
XM_027969885.1 | XP_027825686.1 | 4 - 5 | 956 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
